Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012189025.2 | XP_012044415.1 | 4 - 5 | 537 | 90 | |
XM_027977061.1 | XP_027832862.1 | 4 - 5 | 580 | 90 |