Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161883.1 | NP_001155355.1 | 5 - 6 | 475 | 97 | |
GenBank mRNA | FJ943996.1 | - | 5 - 6 | 475 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |