Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTC4S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970646.1 | XP_027826447.1 | 2 - 3 | 290 | 62 | |
XM_027970647.1 | XP_027826448.1 | 2 - 3 | 234 | 62 | ||
XM_027970648.1 | XP_027826449.1 | 2 - 3 | 218 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |