Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280691.1 | NP_001267620.1 | 1 - 2 | 483 | 65 | |
GenBank mRNA | KC857456.1 | - | 1 - 2 | 483 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |