Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126361.1 | NP_001119833.1 | 4 - 5 | 430 | 65 | |
XM_012174025.2 | XP_012029415.1 | 5 - 6 | 596 | 65 | ||
XM_012174619.3 | XP_012030009.1 | 5 - 6 | 613 | 65 | ||
GenBank mRNA | EU596600.1 | - | 4 - 5 | 430 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |