Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181600.2 | XP_012036990.2 | 3 - 4 | 543 | 118 | |
XM_012181601.2 | XP_012036991.2 | 4 - 5 | 625 | 118 | ||
XM_012181603.2 | XP_012036993.2 | 5 - 6 | 767 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |