Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011230.4 | XP_004011279.1 | 7 - 8 | 1377 | 74 | |
XM_012182674.3 | XP_012038064.1 | 3 - 4 | 1057 | 74 | ||
XM_027972391.1 | XP_027828192.1 | 5 - 6 | 1331 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |