Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190549.3 | XP_012045939.3 | 1 - 2 | 703 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |