Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161860.1 | NP_001155332.1 | 7 - 8 | 532 | 108 | |
GenBank mRNA | FJ943968.1 | - | 7 - 8 | 532 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |