Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164219.1 | NP_001157691.1 | 3 - 4 | 1262 | 85 | |
XM_027963870.1 | XP_027819671.1 | 3 - 4 | 1820 | 85 | ||
XM_027963871.1 | XP_027819672.1 | 3 - 4 | 1818 | 85 | ||
XM_027963872.1 | XP_027819673.1 | 4 - 5 | 1719 | 85 | ||
GenBank mRNA | GQ280389.1 | - | 3 - 4 | 1262 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |