Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958995.1 | XP_027814796.1 | 7 - 8 | 3113 | 80 | |
XM_027958996.1 | XP_027814797.1 | 6 - 7 | 2126 | 80 | ||
XM_027958997.1 | XP_027814798.1 | 6 - 7 | 2038 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |