Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015097.4 | XP_004015146.2 | 11 - 12 | 1374 | 73 | |
XM_012190032.3 | XP_012045422.2 | 11 - 12 | 1313 | 73 |