Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012550.4 | XP_004012599.2 | 9 - 10 | 2376 | 52 | |
XM_012185460.3 | XP_012040850.2 | 9 - 10 | 2333 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |