Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096439.2 | XP_014951925.1 | 3 - 4 | 901 | 112 | |
XM_027970929.1 | XP_027826730.1 | 2 - 3 | 4561 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |