Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096450.2 | XP_014951936.1 | 1 - 2 | 65 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |