Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092470.2 | XP_014947956.2 | 5 - 6 | 2448 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |