Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267876.1 | NP_001254805.1 | 6 - 7 | 685 | 103 | |
XM_027969181.1 | XP_027824982.1 | 6 - 7 | 1327 | 103 | ||
GenBank mRNA | JQ911755.1 | - | 6 - 7 | 685 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |