Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016962.4 | XP_004017011.1 | 2 - 3 | 378 | 112 | |
XM_012096890.3 | XP_011952280.1 | 2 - 3 | 575 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |