Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010473.4 | XP_004010522.2 | 5 - 6 | 1187 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |