Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003164.3 | XP_004003213.1 | 2 - 3 | 257 | 90 | |
XM_027958395.1 | XP_027814196.1 | 2 - 3 | 257 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |