Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103902.2 | XP_014959388.2 | 2 - 3 | 259 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |