Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101087.2 | XP_014956573.2 | 2 - 3 | 457 | 109 | |
XM_027979560.1 | XP_027835361.1 | 2 - 3 | 215 | 109 | ||
XM_027979561.1 | XP_027835362.1 | 3 - 4 | 382 | 109 | ||
XM_027979562.1 | XP_027835363.1 | 2 - 3 | 247 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |