Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968694.1 | XP_027824495.1 | 5 - 6 | 734 | 88 | |
XM_027968695.1 | XP_027824496.1 | 7 - 8 | 870 | 88 | ||
XM_027968696.1 | XP_027824497.1 | 8 - 9 | 927 | 88 | ||
XM_027968697.1 | XP_027824498.1 | 7 - 8 | 865 | 88 | ||
XM_027968698.1 | XP_027824499.1 | 6 - 7 | 777 | 88 | ||
XM_027968699.1 | XP_027824500.1 | 4 - 5 | 656 | 88 | ||
XM_027968700.1 | XP_027824501.1 | 4 - 5 | 541 | 88 | ||
XM_027968702.1 | XP_027824503.1 | 3 - 4 | 424 | 88 |