Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187757.3 | XP_012043147.2 | 2 - 3 | 2385 | 90 | |
XM_012187758.3 | XP_012043148.2 | 2 - 3 | 2656 | 90 | ||
XM_015099366.2 | XP_014954852.2 | 2 - 3 | 1939 | 90 | ||
XM_015099368.2 | XP_014954854.2 | 3 - 4 | 2074 | 90 | ||
XM_027975966.1 | XP_027831767.1 | 2 - 3 | 1874 | 90 | ||
XM_027975967.1 | XP_027831768.1 | 2 - 3 | 3123 | 90 | ||
XM_027975968.1 | XP_027831769.1 | 2 - 3 | 2656 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |