Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184609.3 | XP_012039999.1 | 5 - 6 | 1711 | 87 | |
XM_012184610.2 | XP_012040000.1 | 4 - 5 | 338 | 87 |