Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009363.1 | NP_001009363.1 | 3 - 4 | 132 | 107 | |
GenBank mRNA | GU169085.1 | - | 3 - 4 | 110 | 107 | |
GU169086.1 | - | 3 - 4 | 110 | 107 | ||
GU169087.1 | - | 3 - 4 | 110 | 107 | ||
GU169088.1 | - | 3 - 4 | 110 | 107 | ||
GU169089.1 | - | 3 - 4 | 110 | 107 | ||
X03238.1 | - | 132 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |