Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114763.2 | NP_001108235.1 | 2 - 3 | 577 | 68 | |
GenBank mRNA | EU216424.1 | - | 2 - 3 | 577 | 68 |