Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002035.4 | XP_004002084.1 | 5 - 6 | 990 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |