Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267888.1 | NP_001254817.1 | 4 - 5 | 392 | 74 | |
GenBank mRNA | GU386372.1 | - | 4 - 5 | 547 | 74 | |
GU386373.1 | - | 4 - 5 | 502 | 74 | ||
GU386374.1 | - | 4 - 5 | 392 | 74 | ||
U89874.1 | - | 3 - 4 | 337 | 74 | ||
Z50743.1 | - | 373 | 74 |