Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126364.1 | NP_001119836.1 | 3 - 4 | 397 | 59 | |
XM_027957540.1 | XP_027813341.1 | 4 - 5 | 653 | 59 | ||
XM_027957541.1 | XP_027813342.1 | 4 - 5 | 1901 | 59 | ||
XM_027957542.1 | XP_027813343.1 | 4 - 5 | 492 | 59 | ||
XM_027957543.1 | XP_027813344.1 | 4 - 5 | 499 | 59 | ||
XM_027957544.1 | XP_027813345.1 | 4 - 5 | 664 | 59 | ||
GenBank mRNA | EU596593.1 | - | 3 - 4 | 397 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |