Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009393.1 | NP_001009393.1 | 5 - 6 | 1204 | 85 | |
GenBank mRNA | X68287.1 | - | 1204 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |