Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009395.2 | NP_001009395.1 | 2 - 3 | 278 | 90 | |
XM_012186882.2 | XP_012042272.1 | 3 - 4 | 501 | 90 | ||
XM_027975568.1 | XP_027831369.1 | 5 - 6 | 781 | 90 | ||
XM_027975569.1 | XP_027831370.1 | 4 - 5 | 1141 | 90 | ||
XM_027975570.1 | XP_027831371.1 | 4 - 5 | 610 | 90 | ||
GenBank mRNA | X69561.1 | - | 281 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |