Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009804.1 | NP_001009804.1 | 1 - 2 | 77 | 56 | |
GenBank mRNA | X53037.1 | - | 77 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |