Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161856.1 | NP_001155328.1 | 4 - 5 | 466 | 126 | |
GenBank mRNA | EF564587.1 | - | 3 - 4 | 202 | 126 | |
FJ943986.1 | - | 4 - 5 | 466 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |