Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163055.2 | NP_001156527.1 | 3 - 4 | 424 | 92 | |
GenBank mRNA | EE753586.1 | - | 3 - 4 | 424 | 92 | |
EE767677.1 | - | 3 - 4 | 417 | 92 | ||
FJ958352.1 | - | 2 - 3 | 372 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |