Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205150.1 | NP_001192079.1 | 3 - 4 | 370 | 76 | |
GenBank mRNA | HM017967.1 | - | 3 - 4 | 370 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |