Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139447.1 | NP_001132919.1 | 4 - 5 | 477 | 124 | |
XM_027973929.1 | XP_027829730.1 | 3 - 4 | 382 | 124 | ||
GenBank mRNA | FJ211198.1 | - | 4 - 5 | 477 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |