Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139447.1 | NP_001132919.1 | 7 - 8 | 1027 | 63 | |
XM_027973929.1 | XP_027829730.1 | 6 - 7 | 932 | 63 | ||
GenBank mRNA | FJ211198.1 | - | 7 - 8 | 1027 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |