Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009220.1 | NP_001009220.1 | 2 - 3 | 297 | 70 | |
XM_027966611.1 | XP_027822412.1 | 4 - 5 | 473 | 70 | ||
GenBank mRNA | AF125374.1 | - | 2 - 3 | 297 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |