Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009220.1 | NP_001009220.1 | 7 - 8 | 716 | 59 | |
XM_027966611.1 | XP_027822412.1 | 9 - 10 | 892 | 59 | ||
GenBank mRNA | AF125374.1 | - | 7 - 8 | 716 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |