Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267882.1 | NP_001254811.1 | 2 - 3 | 536 | 62 | |
GenBank mRNA | JQ769145.1 | - | 2 - 3 | 536 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |