Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267882.1 | NP_001254811.1 | 4 - 5 | 865 | 79 | |
GenBank mRNA | JQ769145.1 | - | 4 - 5 | 865 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |