Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OOEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287467.1 | NP_001274396.1 | 2 - 3 | 386 | 105 | |
GenBank mRNA | KF218578.1 | - | 2 - 3 | 386 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |