Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318079.1 | NP_001305008.1 | 3 - 4 | 443 | 87 | |
GenBank mRNA | KR020834.1 | - | 3 - 4 | 443 | 87 |