Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009738.1 | NP_001009738.1 | 3 - 4 | 312 | 88 | |
XM_012101537.3 | XP_011956927.1 | 3 - 4 | 353 | 88 | ||
GenBank mRNA | AF179926.1 | - | 3 - 4 | 312 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |