Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166199.1 | NP_001159671.1 | 3 - 4 | 1040 | 81 | |
XM_012107898.3 | XP_011963288.1 | 3 - 4 | 1075 | 81 | ||
XM_012108258.3 | XP_011963648.1 | 3 - 4 | 1046 | 81 | ||
XM_012108898.3 | XP_011964288.1 | 2 - 3 | 1089 | 81 | ||
GenBank mRNA | GQ221043.1 | - | 3 - 4 | 1040 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |