Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127290.1 | NP_001120762.1 | 2 - 3 | 413 | 87 | |
GenBank mRNA | EU525166.1 | - | 2 - 3 | 261 | 87 | |
EU626221.1 | - | 2 - 3 | 413 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |