Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162568.1 | NP_001156040.1 | 3 - 4 | 426 | 80 | |
XM_012189513.2 | XP_012044903.1 | 3 - 4 | 476 | 80 | ||
GenBank mRNA | FJ958346.1 | - | 3 - 4 | 426 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |