Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126344.1 | NP_001119816.1 | 3 - 4 | 381 | 84 | |
GenBank mRNA | EU579456.1 | - | 3 - 4 | 381 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |