Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126357.1 | NP_001119829.1 | 3 - 4 | 250 | 77 | |
XM_012176231.3 | XP_012031621.1 | 3 - 4 | 410 | 77 | ||
GenBank mRNA | EU596594.1 | - | 3 - 4 | 250 | 77 |